Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COL11A1 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP304904100UL
Description
COL11A1 Polyclonal specifically detects COL11A1 in Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
COL11A1 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
1301 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry | |
Unconjugated | |
alpha-1 polypeptide, COLL6, collagen, type XI, alpha 1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 260-430 of human COL11A1 (NP_542196.2). | |
100 μL | |
Primary | |
Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction