Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COL11A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577919
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577919 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577919 Supplier Invitrogen™ Supplier No. PA577919
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: human COLC11A2-transfected 293T cells (partial fragment) Predicted Reactivity: Mouse (89%), Rat (89%), Bovine (93%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes one of the two alpha chains of type XI collagen, a minor fibrillar collagen. It is located on chromosome 6 very close to but separate from the gene for retinoid X receptor beta. Type XI collagen is a heterotrimer but the third alpha chain is a post-translationally modified alpha 1 type II chain. Proteolytic processing of this type XI chain produces PARP, a proline/arginine-rich protein that is an amino terminal domain. Mutations in this gene are associated with type III Stickler syndrome, otospondylomegaepiphyseal dysplasia (OSMED syndrome), Weissenbacher-Zweymuller syndrome, autosomal dominant non-syndromic sensorineural type 13 deafness (DFNA13), and autosomal recessive non-syndromic sensorineural type 53 deafness (DFNB53). Alternative splicing results in multiple transcript variants. A related pseudogene is located nearby on chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL11A2
Applications Western Blot
Classification Polyclonal
Concentration 0.76 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene COL11A2
Gene Accession No. P13942
Gene Alias COL11A2; Collagen alpha-2(XI) chain; collagen type XI alpha 2; collagen type XI alpha 2 chain; collagen XI-like; collagen, type XI, alpha 2; DAQB-79P13.8; DFNA13; DFNB53; FBCG2; HGNC:2187; HKE5; PARP; pro-a2 chain of collagen type XI; procollagen, type XI, alpha 2; putative collagen XI; STL3; type XI collagen alpha2 chain
Gene Symbols COL11A2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human COL11A2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1302
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.