Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COL12A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552655
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552655 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552655 Supplier Invitrogen™ Supplier No. PA552655
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VTTPPNQRRR TLENLIPDTK YEVSVIPEYF SGPGTPLTGN AATEEVRGNP RDLRVSDPTT STMKLSWSGA PGKVKQYLVT YTPVAGGETQ EVTVRGDTTN TVLQGLKEGT QYALSVTALY ASGAG Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.

This gene encodes the alpha chain of type XII collagen, a member of the FACIT (fibril-associated collagens with interrupted triple helices) collagen family. Type XII collagen is a homotrimer found in association with type I collagen, an association that is thought to modify the interactions between collagen I fibrils and the surrounding matrix. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL12A1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COL12A1
Gene Accession No. Q99715
Gene Alias BA209D8.1; BTHLM2; COL12A1; COL12A1L; collagen alpha-1(XII) chain; collagen type XII alpha 1 chain; collagen type XII proteoglycan; collagen, type XII, alpha 1; DJ234P15.1; UCMD2
Gene Symbols COL12A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COL12A1. Recombinant protein control fragment (Product #RP-89835).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1303
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.