Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COL14A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568472
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568472 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568472 Supplier Invitrogen™ Supplier No. PA568472
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

Type XIV collagen is a fibril-associated collagen with an interrupted triple helix. It interacts with the fibril surface and regulates fibrillogenesis (Ansorge et al., 2009 [PubMed 19136672]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL14A1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene COL14A1
Gene Accession No. Q05707, Q80X19
Gene Alias 5730412L22Rik; AW108078; col14a1; Collagen alpha-1(XIV) chain; collagen type XIV alpha 1 chain; collagen, type XIV, alpha 1; collagen, type XIV, alpha 1 (undulin); procollagen, type XIV, alpha 1; UND; Undulin; undulin (fibronectin-tenascin-related)
Gene Symbols COL14A1
Host Species Rabbit
Immunogen Synthesized peptide derived from human COL14A1 [aa 1571-1620].
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12818, 314981, 7373
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.