Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COL14A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP304388100UL
Description
COL14A1 Polyclonal specifically detects COL14A1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
COL14A1 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
7373 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
collagen alpha-1(XIV) chain, collagen, type XIV, alpha 1, Undulin, undulin (fibronectin-tenascin-related), UNDundulin | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1060-1360 of human COL14A1 (NP_066933.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction