Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COL15A1 Antibody (OTI1C5), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270422AF405
Description
COL15A1 Monoclonal specifically detects COL15A1 in Human samples. It is validated for Western Blot.Specifications
COL15A1 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
COL15A1 | |
Full length recombinant protein of human COL15A1 (NP_001846) produced in HEK293T cells. | |
0.1 mL | |
1306.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI1C5 | |
Western Blot | |
alpha-1 polypeptide, collagen alpha-1(XV) chain, collagen, type XV, alpha 1, FLJ38566 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction