Learn More
Invitrogen™ COL15A1 Polyclonal Antibody

Description
Immunogen sequence: RSSQALAFES SAGIFMGNAG ATGLERFTGS LQQLTVHPDP RTPEELCDPE ESSASGETSG LQEADGVAEI LEAVTYTQAS PKEAKVEPIN TPPTPSSPFE DMELSGEPVP E Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 77%.
Specifications
Specifications
| Antigen | COL15A1 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.20 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | COL15A1 |
| Gene Accession No. | P39059 |
| Gene Alias | COFA1; COL15A1; Collagen alpha-1(XV) chain; collagen type XV alpha 1 chain; collagen type XV proteoglycan; collagen XV, alpha-1 polypeptide; collagen, type XV, alpha 1; endostatin-XV; procollagen type XV; procollagen, type XV; Related to endostatin; Restin; Restin-2; Restin-3; Restin-4; Restin-I; Restin-II; Restin-III; Restin-IV; type XV collagen |
| Gene Symbols | COL15A1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.