Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COL17A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560461
Description
Immunogen sequence: DGTEVTERIV TETVTTRLTS LPPKGGTSNG YAKTASLGGG SRLEKQSLTH GSSGYINSTG STRGHASTSS YRRAHSPAST LPNSPGSTFE RKTHVTR Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
COL17A1 is the alpha chain of type XVII collagen. Unlike most collagens, collagen XVII is a transmembrane protein, and a structural component of hemidesmosomes, multiprotein complexes at the dermal-epidermal basement membrane zone that mediate adhesion of keratinocytes to the underlying membrane. COL17A1 is a member of the FACIT collagen family (fibril-associated collagens with interrupted helices). Moreover, COL17A1 is localized to tissues containing type I collagen so, like other members of this collagen family, it may serve to maintain the integrity of the extracellular matrix. Diseases associated with COL17A1 dysfunction include epithelial recurrent erosion dystrophy and epidermolysis bullosa.
Specifications
COL17A1 | |
Polyclonal | |
Unconjugated | |
COL17A1 | |
120 kDa linear IgA dermatosis antigen; 120 kDa linear IgA disease antigen; 120 kDa linear IgA disease antigen homolog; 180 kDa bullous pemphigoid antigen 2; 97 kDa LAD antigen; 97 kDa linear IgA bullous dermatosis antigen; 97 kDa linear IgA disease antigen; alpha 1 type XVII collagen; BA16H23.2; bA16H23.2 (collagen, type XVII, alpha 1 (BP180)); BP180; BPA-2; Bpag; BPAG2; Bullous pemphigoid antigen 2; bullous pemphigoid antigen 2 (180kD); Col17a1; Collagen alpha-1(XVII) chain; collagen type XVII alpha 1; collagen XVII, alpha-1 polypeptide; collagen, type XVII, alpha 1; ERED; LABD97; LAD-1; Linear IgA bullous disease antigen of 97 kDa; Linear IgA disease antigen 1; procollagen, type XVII, alpha 1; type XVII collagen alpha-1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1308 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UMD9 | |
COL17A1 | |
Recombinant protein corresponding to Human COL17A1. Recombinant protein control fragment (Product #RP-100377). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction