Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COL1A1 Monoclonal Antibody (C1), Invitrogen™

Mouse Monoclonal Antibody
$206.00 - $481.00
Specifications
| Antigen | COL1A1 |
|---|---|
| Clone | C1 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEMA550845
![]() |
Invitrogen™
MA550845 |
200 μL |
Each for $481.00
|
|
|||||
|
FEMA550846
|
Invitrogen™
MA550846 |
20 μL |
Each for $206.00
|
|
|||||
Description
The protein encoded by this gene is the pro-alpha1 chain of type I collagen whose triple helix comprises two alpha1 chains and one alpha2 chain. Type I is a fibril-forming collagen found in most connective tissues and is abundant in bone, cornea, dermis and tendon. Mutations in this gene are associated with osteogenesis imperfecta types I-IV, Ehlers-Danlos syndrome type VIIA, Ehlers-Danlos syndrome Classical type, Caffey Disease and idiopathic osteoporosis. Reciprocal translocations between chromosomes 17 and 22, where this gene and the gene for platelet-derived growth factor beta are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans, resulting from unregulated expression of the growth factor. Two transcripts, resulting from the use of alternate polyadenylation signals, have been identified for this gene.Specifications
| COL1A1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| alpha-1 type 1 collagen; alpha-1 type I collagen; alpha1(I) procollagen; COL1A; Col1a1; Col1a-1; Cola1; Cola-1; COLIA1; Collagen 1; collagen alpha 1 chain type I; collagen alpha-1(I) chain; collagen alpha-1(I) chain preproprotein; Collagen I; collagen of skin, tendon and bone, alpha-1 chain; collagen type I alpha 1; collagen, type 1, alpha 1; collagen, type I, alpha 1; Collagen1A1; EDSC; Mov13; Mov-13; OI1; OI2; OI3; OI4; pro-alpha-1 collagen type 1; procollagen type I, alpha 1; procollagen, type 1, alpha 1; procollagen, type I, alpha 1; Type I Collagen; type I proalpha 1; type I procollagen alpha 1 chain; type I-alpha 1 collagen | |
| COL1A1 | |
| IgG1 κ | |
| Protein A/G | |
| COL1A1 |
| C1 | |
| Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Pig | |
| P02452 | |
| 100738123, 1277 | |
| Recombinant Human COL1a1 protein, Arg253-Ala471 (Accession #P02452). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title