Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COL1A2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP157987
Description
COL1A2 Polyclonal specifically detects COL1A2 in Human, Mouse samples. It is validated for Western Blot.Specifications
COL1A2 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
alpha 2(I)-collagen, Alpha-2 type I collagen, collagen alpha-2(I) chain, collagen I, alpha-2 polypeptide, collagen of skin, tendon and bone, alpha-2 chain, collagen, type I, alpha 2, OI4, osteogenesis imperfecta type IV, type I procollagen | |
Rabbit | |
150 kDa | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%; Pig: 92%; Rabbit: 85%; Rat: 85%; Chicken: 84%; Xenopus: 78%; Guinea pig: 78%; Mouse: 78%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Goat, Rabbit, Zebrafish | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
P08123 | |
COL1A2 | |
Synthetic peptides corresponding to COL1A2(collagen, type I, alpha 2) The peptide sequence was selected from the middle region of COL1A2. Peptide sequence PGSVGPAGPRGPAGPSGPAGKDGRTGHPGTVGPAGIRGPQGHQGPAGPPG. | |
Affinity purified | |
RUO | |
1278 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction