Learn More
Invitrogen™ COL20A1 Monoclonal Antibody (1F5)

Description
Peptide Sequence: MRGLEGTAGL PGPPGPRGFQ GMAGARGTSG ERGPPGTVGP TGLPGPKGER GEKGEPQSLA TLYQLVSQAC ESAIQTHVSK FDSFHENTRP PMPILEQKLE PGTEPLGSPG TRSKALVPGE WGRGGRHLEG RGEPGAVGQM GSPGQQGAST QGLWE.
Specifications
Specifications
| Antigen | COL20A1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 1F5 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | COL20A1 |
| Gene Accession No. | Q9P218 |
| Gene Alias | 1700051I12Rik; bA261N11.4; Col20a1; Collagen alpha-1(XX) chain; collagen type XX alpha 1 chain; collagen, type XX, alpha 1; collagen-like protein; KIAA1510; RGD1563901 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.