Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COL23A1 Monoclonal Antibody (468642)
GREENER_CHOICE

Catalog No. PIMA524188
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524188 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524188 Supplier Invitrogen™ Supplier No. MA524188
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

In direct ELISAs and Western blots, 50% cross-reactivity with recombinant human Collagen XXV alpha 1 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

COL23A1 is a member of the transmembrane collagens, a subfamily of the nonfibrillar collagens that contain a single pass hydrophobic transmembrane domain (Banyard et al., 2003 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL23A1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 468642
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene COL23A1
Gene Accession No. Q86Y22
Gene Alias 2810458L13Rik; AI429655; Col23a1; Collagen alpha-1(XXIII) chain; collagen type XXIII alpha 1 chain; collagen XXIII; collagen, type XXIII, alpha 1; DKFZp434K0621; procollagen type XXIII alpha 1; procollagen, type XXIII, alpha 1
Gene Symbols COL23A1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Collagen XXIII alpha 1 Glu111-Lys540.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91522
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.