Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COL23A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100447
Description
Antibody detects endogenous levels of total Collagen Type XXIII.
COL23A1 is a member of the transmembrane collagens, a subfamily of the nonfibrillar collagens that contain a single pass hydrophobic transmembrane domain (Banyard et al., 2003 ).
Specifications
COL23A1 | |
Polyclonal | |
Unconjugated | |
COL23A1 | |
2810458L13Rik; AI429655; Col23a1; Collagen alpha-1(XXIII) chain; collagen type XXIII alpha 1 chain; collagen XXIII; collagen, type XXIII, alpha 1; DKFZp434K0621; procollagen type XXIII alpha 1; procollagen, type XXIII, alpha 1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
237759, 353303, 91522 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q810Y4, Q86Y22, Q8K4G2 | |
COL23A1 | |
A synthesized peptide derived from human COL23A1(Accession Q86Y22), corresponding to amino acid residues P120-D170. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction