Learn More
Invitrogen™ COL24A1 Monoclonal Antibody (1D8)

Description
Peptide Sequence: PRWTSTQTSG PGLPIGFKGW NGQIFKVNTL LEPKVLSDDC KIQDGSWHKA TFLFHTQEPN QLPVIEVQKL PHLKTERKYY IDSSSVCFL.
Specifications
Specifications
| Antigen | COL24A1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1D8 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | COL24A1 |
| Gene Accession No. | Q17RW2 |
| Gene Alias | COL24A1; Collagen alpha-1(XXIV) chain; collagen type XXIV alpha 1; collagen, type XXIV, alpha 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.