Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COL26A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582951
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582951 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582951 Supplier Invitrogen™ Supplier No. PA582951
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ANLVSYRTLI RPTYRVSYRT VTVLEWRCCP GFTGSNCDEE CMNCTRLSDM SERLTTLEAK VLLLEAAERP SSPDNDLPAP ESTPPTWNED FLPDAIPLAH PVP Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 75%.

COL26A1 (EMID2) is a 441 amino acid protein encoded by a gene mapping to human chromosome 7. Chromosome 7 is approximately 158 milllion bases long, encodes over 1,000 genes and makes up about 5% of the human genome. Chromosome 7 has been linked to osteogenesis imperfecta, Pendred syndrome, lissencephaly, citrullinemia and Shwachman-Diamond syndrome. The deletion of a portion of the q arm of chromosome 7 is associated with Williams-Beuren syndrome, a condition characterized by mild mental retardation, an unusual comfort and friendliness with strangers, and an elfin appearance. Deletions of portions of the q arm of chromosome 7 are also seen in a number of myeloid disorders including cases of acute myelogenous leukemia and myelodysplasia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL26A1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COL26A1
Gene Accession No. Q96A83
Gene Alias 9430032K24Rik; alpha-1 type XXVI collagen; BC002218; Col26a; COL26A1; Collagen alpha-1(XXVI) chain; collagen type XXVI alpha 1; Collagen XXVI; collagen, type XXVI, alpha 1; EMI domain containing 2; EMI domain-containing protein 2; EMI6; EMID2; emilin and multimerin domain-containing protein 2; EMU2; procollagen, type XXVI, alpha 1; SH2B
Gene Symbols COL26A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COL26A1. Recombinant protein control fragment (Product #RP-108147).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 136227
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.