Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COL28A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583683
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583683 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583683 Supplier Invitrogen™ Supplier No. PA583683
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SESLSVTRDQ DEDDKAPEPT WADDLPATTS SEATTTPRPL LSTPVDGAED PRCLEALKPG NCGEYVVRWY YDKQVNSCAR FWFSGCNGSG NRFNSEKECQ ET Highest antigen sequence indentity to the following orthologs: Mouse - 57%, Rat - 58%.

COL28A1 belongs to a class of collagens containing von Willebrand factor (VWF; MIM 613160) type A (VWFA) domains (Veit et al., 2006 [PubMed 16330543]).[supplied by OMIM, Nov 2010].
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL28A1
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COL28A1
Gene Accession No. Q2UY09
Gene Alias COL28; COL28A1; Collagen alpha-1(XXVIII) chain; collagen type XXVIII alpha 1 chain; collagen, type XXVIII, alpha 1
Gene Symbols COL28A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COL28A1. Recombinant protein control fragment (Product #RP-100347).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340267
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.