Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COL4A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109568
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109568 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109568 Supplier Invitrogen™ Supplier No. PA5109568
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

COL4A2 Polyclonal Antibody for ELISA, Western Blot

Collagen IV is a major constituent of the basement membranes along with laminins and enactins. Collagen IV is composed of alpha 1 IV chain and alpha 2 IV chain in 2:1 ratio. Collagen IV can form insoluble fibers with high tensile strength. Antibody to collagen IV is useful in detecting the loss of parts of basement membrane in carcinomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL4A2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene COL4A2
Gene Accession No. P08122, P08572
Gene Alias alpha-2 collagen type IV; canstatin; COL4A2; Col4a-2; Collagen alpha-2(IV) chain; collagen type IV alpha 2; collagen, type IV, alpha 2; ICH; POREN2; procollagen, type IV, alpha 2
Gene Symbols COL4A2
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12827, 1284, 306628
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.