Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COL4A3BP Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002114AF405
Description
COL4A3BP Polyclonal antibody specifically detects COL4A3BP in Human samples. It is validated for Western BlotSpecifications
COL4A3BP | |
Polyclonal | |
Western Blot | |
Ceramide transfer protein, CERTFLJ20597, CERTL, collagen type IV alpha-3-binding protein, collagen, type IV, alpha 3 (Goodpasture antigen) binding protein, Goodpasture antigen-binding protein, GPBPSTARD11ceramide transporter, hCERT, lipid-transfer protein CERTL, StARD11, StAR-related lipid transfer (START) domain containing 11, StAR-related lipid transfer protein 11, START domain containing 11, START domain-containing protein 11 | |
The immunogen recognized by this antibody maps to a region between residue 300 and 350 of human CERTL(GPBP) using the numbering given in entry NP_005704.1 (GeneID 10087). | |
0.1 mL | |
Cell Biology, Cellular Markers, Immune System Diseases, Immunology | |
10087 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction