Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COL4A3BP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP215943
Description
COL4A3BP Polyclonal specifically detects COL4A3BP in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
COL4A3BP | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
Ceramide transfer protein, CERTFLJ20597, CERTL, collagen type IV alpha-3-binding protein, collagen, type IV, alpha 3 (Goodpasture antigen) binding protein, Goodpasture antigen-binding protein, GPBPSTARD11ceramide transporter, hCERT, lipid-transfer protein CERTL, StARD11, StAR-related lipid transfer (START) domain containing 11, StAR-related lipid transfer protein 11, START domain containing 11, START domain-containing protein 11 | |
Rabbit | |
71 kDa | |
0.1 mL | |
Cytoskeleton Markers, Immune System Diseases, Immunology | |
10087 | |
Human | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.89mg/mL | |
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
Q9Y5P4 | |
COL4A3BP | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human COL4A3BP. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human. Expected to cross react based on sequence identity: Mouse (100%), Rat (100%), Xenopus laevis (100%), Bovine (100%), Xenopus tropicalis (100%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction