Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COL4A3BP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117302
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117302 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117302 Supplier Invitrogen™ Supplier No. PA5117302
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Centrifuge briefly prior to opening.

CERT (GPBP) is a GC-rich promotor-specific transactivating transcription factor. This protein is suggested to play a role in the development of atherosclerosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL4A3BP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene CERT1
Gene Accession No. Q9Y5P4
Gene Alias 2810404O15Rik; 9230101K08Rik; alpha 3 type IV collagen binding protein; AU016711; Ceramide transfer protein; ceramide transport protein; ceramide transporter; Cert; CERTL; Col4a3bp; collagen type IV alpha 3 binding protein; collagen type IV alpha-3-binding protein; collagen, type IV, alpha 3 (Goodpasture antigen) binding protein; goodpasture antigen-binding protein; GPBP; hCERT; lipid-transfer protein CERTL; MRD34; procollagen, type IV, alpha 3 (Goodpasture antigen) binding protein; STARD11; StAR-related lipid transfer (START) domain containing 11; stAR-related lipid transfer protein 11; START domain-containing protein 11
Gene Symbols CERT1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human COL4A3BP. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10087
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.