Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COL4A4 Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | COL4A4 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119041
|
Invitrogen™
PA5119041 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5122082
|
Invitrogen™
PA5122082 |
20 μL |
Each for $209.50
|
|
|||||
Description
COL4A4 Polyclonal Antibody for Western Blot, ICC/IF
Collagens are highly conserved and are characterized by an uninterrupted Glycine-X-Y triplet repeat. Col1, the fibrillar collagen found in most connective tissues, is the only component of the collagen found in cartilage. Mutations in this gene are associated with osteogenesis imperfecta, Ehlers-Danlos syndrome, and idiopathic osteoporosis. Col2, a fibrillar collagen found in cartilage and the vitreous humor of the eye. Mutations in this gene are associated with achondrogenesis, chondrodysplasia, early onset familial osteoarthritis, SED congenita, Langer-Saldino achondrogenesis, Kniest dysplasia. Col3 is a fibrillar collagen that is found in extensible connective tissues such as skin, lung, and the vascular system, frequently in association with type I collagen. Mutations in this gene are associated with Ehlers-Danlos syndrome type IV, aortic and arterial aneurysms. Col4 is a major consitiuent of the basement membranes along with laminins and enactins. It is composed of alpha 1 IV chain and alpha 2 IV chain in 2:1 ratio. Col4 antibodies are useful in detecting the loss of parts of basement membrane in carcinomas. Col5 is found in tissues containing Col1 and appears to regulate the assembly of heterotypic fibers composed of both Col1 and Col5. Col5 is closely related to type XI collagen; mutations in this gene are associated with Ehlers-Danlos syndrome.Specifications
COL4A4 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
COL4A4 | |
CA44; COL4A4; Collagen 4; Collagen alpha-4(IV) chain; Collagen IV, alpha-4 polypeptide; collagen of basement membrane, alpha-4 chain; collagen type IV alpha 4 chain; collagen, type IV, alpha 4 | |
COL4A4 | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
P53420 | |
1286 | |
Recombinant protein Collagen Type IV Alpha 4. The antigen corresponds to amino acid range 1469-1690 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title