Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COL4A5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121239
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121239 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121239 Supplier Invitrogen™ Supplier No. PA5121239
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: 22Rv1, NCI-H460, HepG2. The target is usually found in the following locations: Secreted, basement membrane, extracellular matrix, extracellular space. Immunogen sequence: SSVAHGFLIT RHSQTTDAPQ CPQGTLQVYE GFSLLYVQGN KRAHGQDLGT AGSCLRRFST MPFMFCNINN VCNFASRNDY SYWLSTPEPM PMSMQPLKGQ SIQPFISRCA VCEAPAVVIA VHSQTIQIPH CPQGWDSLWI GYSFMMHTSA GAEGSGQALA SPGSCLEEFR SAPFIECHGR GTCNYYANSY SFWLATVDVS DMFSKPQSET LKAGDLRTRI SRCQVCMKRT.

Collagens are highly conserved and are characterized by an uninterrupted Glycine-X-Y triplet repeat. Col1, the fibrillar collagen found in most connective tissues, is the only component of the collagen found in cartilage. Mutations in this gene are associated with osteogenesis imperfecta, Ehlers-Danlos syndrome, and idiopathic osteoporosis. Col2, a fibrillar collagen found in cartilage and the vitreous humor of the eye. Mutations in this gene are associated with achondrogenesis, chondrodysplasia, early onset familial osteoarthritis, SED congenita, Langer-Saldino achondrogenesis, Kniest dysplasia. Col3 is a fibrillar collagen that is found in extensible connective tissues such as skin, lung, and the vascular system, frequently in association with type I collagen. Mutations in this gene are associated with Ehlers-Danlos syndrome type IV, aortic and arterial aneurysms. Col4 is a major consitiuent of the basement membranes along with laminins and enactins. It is composed of alpha 1 IV chain and alpha 2 IV chain in 2:1 ratio. Col4 antibodies are useful in detecting the loss of parts of basement membrane in carcinomas. Col5 is found in tissues containing Col1 and appears to regulate the assembly of heterotypic fibers composed of both Col1 and Col5. Col5 is closely related to type XI collagen; mutations in this gene are associated with Ehlers-Danlos syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL4A5
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene COL4A5
Gene Accession No. P29400
Gene Alias ASLN; ATS; CA54; CO4A5; COL4A5; Collagen alpha-5(IV) chain; collagen IV, alpha-5 polypeptide; collagen of basement membrane, alpha-5 chain; collagen type IV alpha 5 chain; collagen type IV alpha5 chain; collagen, type IV, alpha 5; dA149D17.3; dA24A23.1; procollagen type IV alpha 5; procollagen, type IV, alpha 5
Gene Symbols COL4A5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1462-1691 of human COL4A5 (NP_203699.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1287
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.