Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COL4A6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584691
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584691 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584691 Supplier Invitrogen™ Supplier No. PA584691
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EPILSTIQGM PGDRGDSGSQ GFRGVIGEPG KDGV Highest antigen sequence indentity to the following orthologs: Mouse - 53%, Rat - 53%.

Type I collagen, the fibrillar collagen found in most connective tissues, and the only component of the collagen found in cartilage. Mutations in this gene are associated with osteogenesis imperfecta, Ehlers-Danlos syndrome, and idiopathic osteoporosis. Reciprocal translocations between chromosomes 17 and 22, where this gene and the gene for platelet-derived growth factor beta are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans, resulting from unregulated expression of the growth factor. Two transcripts, resulting from the use of alternate polyadenylation signals, have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL4A6
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COL4A6
Gene Accession No. Q14031
Gene Alias BB116301; Col4a6; Collagen 4; Collagen alpha-6(IV) chain; collagen IV, alpha-6 polypeptide; collagen of basement membrane, alpha-6; collagen type IV alpha 6 chain; collagen, type IV, alpha 6; CXDELq22.3; DELXq22.3; DFNX6; dJ889N15.4 (Collagen Alpha 6(IV)); procollagen type IV alpha 6; procollagen, type IV, alpha 6
Gene Symbols COL4A6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COL4A6. Recombinant protein control fragment (Product #RP-107427).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1288
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.