Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COL5A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583158
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583158 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583158 Supplier Invitrogen™ Supplier No. PA583158
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PVEAAKETTE VPEELTPTPT EAAPMPETSE GAGKEEDVGI GDYDYVPSED YYTPSPYDDL TYGEGEENPD QPTDPGAGAE IPTSTADTSN SSNPAPPPGE GADDLEGEFT EETIRNLDEN YYDPYYDPTS SPSEIGPGMP ANQDTIYE Highest antigen sequence indentity to the following orthologs: Mouse - 72%, Rat - 76%.

Type I collagen, the fibrillar collagen found in most connective tissues, and the only component of the collagen found in cartilage. Mutations in this gene are associated with osteogenesis imperfecta, Ehlers-Danlos syndrome, and idiopathic osteoporosis. Reciprocal translocations between chromosomes 17 and 22, where this gene and the gene for platelet-derived growth factor beta are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans, resulting from unregulated expression of the growth factor. Two transcripts, resulting from the use of alternate polyadenylation signals, have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL5A1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COL5A1
Gene Accession No. P20908
Gene Alias AI413331; alpha 1 (V) collagen; alpha1(V) collagen; cha1V; COL5A1; collagen alpha 1 (V); collagen alpha-1(V) chain; collagen type V alpha 1; collagen type V alpha 1 chain; collagen, type V, alpha 1; EDSC; pro-alpha1(V) collagen; procollagen type V alpha 1; procollagen, type V, alpha 1; type V collagen; type V collagen, alpha 1 chain, NH2-terminal domain
Gene Symbols COL5A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COL5A1. Recombinant protein control fragment (Product #RP-102573).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1289
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.