Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COL5A2 Monoclonal Antibody (C4), Invitrogen™

Catalog No. PIMA543882
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543882 20 μL
PIMA543881 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543882 Supplier Invitrogen™ Supplier No. MA543882
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

COL5A2 is an alpha chain for one of the low abundance fibrillar collagens. Fibrillar collagen molecules are trimers that can be composed of one or more types of alpha chains. Type V collagen is found in tissues containing type I collagen and appears to regulate the assembly of heterotypic fibers composed of both type I and type V collagen. This protein is closely related to type XI collagen and it is possible that the collagen chains of types V and XI constitute a single collagen type with tissue-specific chain combinations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL5A2
Applications Western Blot
Classification Monoclonal
Clone C4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene COL5A2
Gene Accession No. P05997
Gene Alias 1110014L14Rik; AB collagen; col5a2; Collagen alpha-2(V) chain; collagen type V alpha 2 chain; collagen, fetal membrane, A polypeptide; collagen, type V, alpha 2; D230017N05; EDSC; procollagen, type V, alpha 2; type V preprocollagen alpha 2 chain
Gene Symbols COL5A2
Host Species Mouse
Immunogen Cys82-Glu308 (Accession P51861), with N-terminal His and GST Tag.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1290
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.