Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COL5A2 Polyclonal Antibody

COL5A2 Polyclonal Antibody
Supplier: Thermo Scientific PA575809
Description
COL5A2 Polyclonal Antibody for Western Blot
Specifications
| COL5A2 | |
| Polyclonal | |
| Unconjugated | |
| COL5A2 | |
| 1110014L14Rik; AB collagen; col5a2; Collagen alpha-2(V) chain; collagen type V alpha 2 chain; collagen, fetal membrane, A polypeptide; collagen, type V, alpha 2; D230017N05; EDSC; procollagen, type V, alpha 2; type V preprocollagen alpha 2 chain | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12832, 1290, 85250 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P05997, Q3U962 | |
| COL5A2 | |
| Synthetic peptide corresponding to residues in Human COL5A2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction