Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COL5A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA514245
Description
COL5A2 Polyclonal Antibody for Western Blot, Flow
COL5A2 is an alpha chain for one of the low abundance fibrillar collagens. Fibrillar collagen molecules are trimers that can be composed of one or more types of alpha chains. Type V collagen is found in tissues containing type I collagen and appears to regulate the assembly of heterotypic fibers composed of both type I and type V collagen. This protein is closely related to type XI collagen and it is possible that the collagen chains of types V and XI constitute a single collagen type with tissue-specific chain combinations.
Specifications
| COL5A2 | |
| Polyclonal | |
| Unconjugated | |
| COL5A2 | |
| 1110014L14Rik; AB collagen; col5a2; Collagen alpha-2(V) chain; collagen type V alpha 2 chain; collagen, fetal membrane, A polypeptide; collagen, type V, alpha 2; D230017N05; EDSC; procollagen, type V, alpha 2; type V preprocollagen alpha 2 chain | |
| Rabbit | |
| Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
| RUO | |
| 12832, 1290 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P05997, Q3U962 | |
| COL5A2 | |
| KLH conjugated synthetic peptide between 90-118 amino acids from the N-terminal region of human COL5A2. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction