Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COL5A3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144797
Description
Immunogen sequence: STDIPKTETP APNLPPTPTP LVVTSTVTTG LNATILEGSL DPDSGTELGT LETKAAREDE EGDDSTMGPD FRAAEYPSRT QFQ Antigen sequence identity: Mouse 52%; Rat 55%.
Type I collagen, the fibrillar collagen found in most connective tissues, and the only component of the collagen found in cartilage. Mutations in this gene are associated with osteogenesis imperfecta, Ehlers-Danlos syndrome, and idiopathic osteoporosis. Reciprocal translocations between chromosomes 17 and 22, where this gene and the gene for platelet-derived growth factor beta are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans, resulting from unregulated expression of the growth factor. Two transcripts, resulting from the use of alternate polyadenylation signals, have been identified for this gene.
Specifications
COL5A3 | |
Polyclonal | |
Unconjugated | |
COL5A3 | |
COL5A3; Collagen 5; Collagen alpha-3(V) chain; collagen type V alpha 3; collagen, type V, alpha 3; collagen, type V, alpha 4; pro-(alpha)3(V) collagen; Pro-alpha3(V); procollagen, type V, alpha 3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
50509 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P25940 | |
COL5A3 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 285-367. Recombinant protein control fragment (Product #RP-98883). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction