Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COL6A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583085
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583085 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583085 Supplier Invitrogen™ Supplier No. PA583085
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VKVFSVAITP DHLEPRLSII ATDHTYRRNF TAADWGQSRD AEEAISQTID TIVDMIKNNV EQVCCSFECQ PAR Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 93%.

The extensive family of COL gene products (collagens) is composed of several chain types, including fibril-forming interstitial collagens (typesI, II, III and V) and basement membrane collagens (type IV), each type containing multiple isoforms. Collagens are fibrous, extracellular matrix proteins with high tensile strength and are the major components of connective tissue, such as tendons and cartilage. All collagens contain a triple helix domain and frequently show lateral self-association in order to form complex connective tissues. Several collagens also play a role in cell adhesion, important for maintaining normal tissue architecture and function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL6A1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Col6a1
Gene Accession No. P12109
Gene Alias AI747156; alpha 1 (VI) chain (61 AA); BTHLM1; Col6a1; Col6a-1; Collagen alpha-1(VI) chain; collagen type VI alpha 1; collagen type VI alpha 1 chain; collagen VI, alpha-1 polypeptide; collagen, type VI, alpha 1; OPLL; procollagen, type VI, alpha 1; RGD1565398; UCHMD1
Gene Symbols Col6a1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COL6A1. Recombinant protein control fragment (Product #RP-95886).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1291
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.