Learn More
Invitrogen™ COL6A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121228
Description
Positive test controls include: SKOV3, HepG2, HT-1080, Mouse heart, Mouse lung, Mouse pancreas. The target is usually found in the following locations: Secreted, extracellular matrix, extracellular space. Immunogen sequence: AQDEPETPRA VAFQDCPVDL FFVLDTSESV ALRLKPYGAL VDKVKSFTKR FIDNLRDRYY RCDRNLVWNA GALHYSDEVE IIQGLTRMPG GRDALKSSVD AVKYFGKGTY TDCAIKKGLE QLLVGGSHLK ENKYLIVVTD GHPLEGYKEP CGGLEDAVNE AKHLGVKVFS VAITPDHLEP RLSIIATDHT YRRNFTAADW GQSRDAEEAI SQTIDTIVDM IKNNVEQVCC SF.
The extensive family of COL gene products (collagens) is composed of several chain types, including fibril-forming interstitial collagens (typesI, II, III and V) and basement membrane collagens (type IV), each type containing multiple isoforms. Collagens are fibrous, extracellular matrix proteins with high tensile strength and are the major components of connective tissue, such as tendons and cartilage. All collagens contain a triple helix domain and frequently show lateral self-association in order to form complex connective tissues. Several collagens also play a role in cell adhesion, important for maintaining normal tissue architecture and function.
Specifications
| COL6A1 | |
| Polyclonal | |
| Unconjugated | |
| Col6a1 | |
| AI747156; alpha 1 (VI) chain (61 AA); BTHLM1; Col6a1; Col6a-1; Collagen alpha-1(VI) chain; collagen type VI alpha 1; collagen type VI alpha 1 chain; collagen VI, alpha-1 polypeptide; collagen, type VI, alpha 1; OPLL; procollagen, type VI, alpha 1; RGD1565398; UCHMD1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 12833, 1291 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.37 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P12109, Q04857 | |
| Col6a1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 19-250 of human Collagen VI/Collagen VI/COL6A1 (NP_0018392). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.