Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COL6A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565085
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565085 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565085 Supplier Invitrogen™ Supplier No. PA565085
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KGTVHFAVVIT DGHVTGSPCG GIKLQAERAR EEGIRLFAVA PNQNLKEQGL RDIASTPHEL YRNDYATMLP DSTEIDQDTI NRIIKVMKHE AYGECYKVSC Highest antigen sequence identity to the following orthologs - mouse 91%, rat 90%.

This gene encodes one of the three alpha chains of type VI collagen, a beaded filament collagen found in most connective tissues. The product of this gene contains several domains similar to von Willebrand Factor type A domains. These domains have been shown to bind extracellular matrix proteins, an interaction that explains the importance of this collagen in organizing matrix components. Mutations in this gene are associated with Bethlem myopathy and Ullrich scleroatonic muscular dystrophy. Three transcript variants have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL6A2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COL6A2
Gene Accession No. P12110
Gene Alias BTHLM1; Col6a2; Col6a-2; Collagen alpha-2(VI) chain; collagen type VI alpha 2; collagen VI, alpha-2 polypeptide; collagen, type VI, alpha 2; human mRNA for collagen VI alpha-2 C-terminal globular domain; PP3610; procollagen, type VI, alpha 2; UCMD1
Gene Symbols COL6A2
Host Species Rabbit
Immunogen Recombinant Human COL6A2. Recombinant protein control fragment (Product #RP-104621).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1292
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.