Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COL6A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143897
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143897 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143897 Supplier Invitrogen™ Supplier No. PA5143897
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human HCCT tissue, human placenta tissue. IHC: human colonic adenocarcinoma tissue, human gastric adenocarcinoma tissue, human liver cancer tissue, human liver cancer tissue, human liver cancer tissue, human lymphadenoma tissue, human placenta tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

This gene encodes one of the three alpha chains of type VI collagen, a beaded filament collagen found in most connective tissues. The product of this gene contains several domains similar to von Willebrand Factor type A domains. These domains have been shown to bind extracellular matrix proteins, an interaction that explains the importance of this collagen in organizing matrix components. Mutations in this gene are associated with Bethlem myopathy and Ullrich scleroatonic muscular dystrophy. Three transcript variants have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL6A2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene COL6A2
Gene Accession No. P12110
Gene Alias BTHLM1; Col6a2; Col6a-2; Collagen alpha-2(VI) chain; collagen type VI alpha 2; collagen VI, alpha-2 polypeptide; collagen, type VI, alpha 2; human mRNA for collagen VI alpha-2 C-terminal globular domain; PP3610; procollagen, type VI, alpha 2; UCMD1
Gene Symbols COL6A2
Host Species Rabbit
Immunogen E.coli-derived human Collagen VI/COL6A2 recombinant protein (Position: N35-D949).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1292
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.