Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COL6A3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112163
Description
This product can be used in direct ELISA.
This gene encodes the alpha-3 chain, one of the three alpha chains of type VI collagen, a beaded filament collagen found in most connective tissues. The alpha-3 chain of type VI collagen is much larger than the alpha-1 and -2 chains. This difference in size is largely due to an increase in the number of subdomains, similar to von Willebrand Factor type A domains, that are found in the amino terminal globular domain of all the alpha chains. These domains have been shown to bind extracellular matrix proteins, an interaction that explains the importance of this collagen in organizing matrix components. Mutations in the type VI collagen genes are associated with Bethlem myopathy, a rare autosomal dominant proximal myopathy with early childhood onset. Mutations in this gene are also a cause of Ullrich congenital muscular dystrophy, also referred to as Ullrich scleroatonic muscular dystrophy, an autosomal recessive congenital myopathy that is more severe than Bethlem myopathy. Multiple transcript variants have been identified, but the full-length nature of only some of these variants has been described. [provided by RefSeq, Jun 2009].
Specifications
COL6A3 | |
Polyclonal | |
PBS with no preservative | |
P12111 | |
COL6A3 | |
Recombinant Human COL6A3/Collagen-VI protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Western Blot | |
Unconjugated | |
COL6A3 | |
AI507288; BTHLM1; CO6A3; COL6A3; Col6a-3; collagen alpha 3 chain type VI; collagen alpha3(VI); collagen alpha-3(VI) chain; collagen type VI alpha 3 chain; collagen VI, alpha-3 polypeptide; collagen, type VI, alpha 3; DYT27; procollagen, type VI, alpha 3; type VI collagen alpha 3 subunit; UCMD1 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
1293 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction