Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COL6A5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560254
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560254 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560254 Supplier Invitrogen™ Supplier No. PA560254
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDVVFLIDNS RNIAKDEFKA VKALVSSVID NFNIASDPLI SDSGDRIALL SYSPWESSRR KMGTVKTEFD FITYDNQLLM KNHIQTSFQQ LNGEATIGRA L Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 69%. Immunohistochemistry retrieval method: HIER pH 6.

This gene encodes a member of the collagen superfamily of proteins. The encoded protein contains multiple von Willebrand factor A-like domains and may interact with the alpha 1 and alpha 2 chains of collagen VI to form the complete collagen VI trimer. Polymorphisms in this gene may be linked to dermal phenotypes, such as eczema. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL6A5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COL6A5
Gene Accession No. A8TX70
Gene Alias Col29a1; Col6a5; Collagen alpha-1(XXIX) chain; collagen alpha-5(VI) chain; collagen type VI alpha 5; collagen type VI alpha 5 chain; collagen, type VI, alpha 5; collagen, type XXIX, alpha 1; EG665033; Gm7455; RGD1565804; von Willebrand factor A domain-containing protein 4; VWA4
Gene Symbols COL6A5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COL6A5. Recombinant protein control fragment (Product #a href=.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 256076
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.