Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ COL7A1 Monoclonal Antibody (C7)
Mouse Monoclonal Antibody
$243.00 - $522.50
Specifications
Antigen | COL7A1 |
---|---|
Clone | C7 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541570
|
Invitrogen™
MA541570 |
200 μL |
Each for $522.50
|
|
|||||
PIMA543880
|
Invitrogen™
MA543880 |
20 μL |
Each for $243.00
|
|
|||||
Description
COL7A1 Monoclonal Antibody for Western Blot
This gene encodes the alpha chain of type VII collagen. The type VII collagen fibril, composed of three identical alpha collagen chains, is restricted to the basement zone beneath stratified squamous epithelia. It functions as an anchoring fibril between the external epithelia and the underlying stroma. Mutations in this gene are associated with all forms of dystrophic epidermolysis bullosa. In the absence of mutations, however, an acquired form of this disease can result from an autoimmune response made to type VII collagen. [provided by RefSeq, Jul 2008].Specifications
COL7A1 | |
1 mg/mL | |
Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q02388 | |
1294 | |
Recombinant protein Collagen Type VII. The antigen corresponds to amino acid range 190-472 of the target protein. | |
Antibody |
C7 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG1 κ | |
Human | |
COL7A1 | |
alpha-1 type VII collagen; AW209154; COL7A1; collagen alpha-1(VII) chain; collagen type VII alpha 1; collagen type VII alpha 1 chain; collagen vii; collagen VII, alpha-1 polypeptide; collagen, type VII, alpha 1; EBD1; EBDCT; EBR1; HGNC:2214; LC collagen; long chain collagen; long-chain collagen; NDNC8; procollagen, type VII, alpha 1 | |
COL7A1 | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title