Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COL8A1 Monoclonal Antibody (C3)

Catalog No. PIMA543879
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543879 20 μL
PIMA541571 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543879 Supplier Invitrogen™ Supplier No. MA543879
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

COL8A1 Monoclonal Antibody for Western Blot

The extensive family of COL gene products (collagens) is composed of several chain types, including fibril-forming interstitial collagens (types I, II, III and V) and basement membrane collagens (type IV), each type containing multiple isoforms. Collagens are fibrous, extracellular matrix proteins with high tensile strength and are the major components of connective tissue, such as tendons and cartilage. All collagens contain a triple helix domain and frequently show lateral self-association in order to form complex connective tissues. Several collagens also play a role in cell adhesion, important for maintaining normal tissue architecture and function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL8A1
Applications Western Blot
Classification Monoclonal
Clone C3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene COL8A1
Gene Accession No. P27658, Q00780
Gene Alias C3orf7; cell proliferation-inducing protein 41; Col8a1; Col8a-1; Collagen alpha-1(VIII) chain; collagen type VIII alpha 1; collagen VIII, alpha-1 polypeptide; collagen, type VIII, alpha 1; endothelial collagen; procollagen type VIII alpha 1; procollagen, type VIII, alpha 1; smag-64; smooth muscle cell-expressed and macrophage conditioned medium-induced protein smag-64; Vastatin
Gene Symbols COL8A1
Host Species Mouse
Immunogen Recombinant protein Collagen Type VIII Alpha 1. The antigen corresponds to amino acid range 575-743 of the target protein.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12837, 1295
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.