Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COL8A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597604
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597604 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597604 Supplier Invitrogen™ Supplier No. PA597604
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

COL8A1 Polyclonal Antibody for Western Blot, IHC (P), ICC/IF, ELISA

The extensive family of COL gene products (collagens) is composed of several chain types, including fibril-forming interstitial collagens (types I, II, III and V) and basement membrane collagens (type IV), each type containing multiple isoforms. Collagens are fibrous, extracellular matrix proteins with high tensile strength and are the major components of connective tissue, such as tendons and cartilage. All collagens contain a triple helix domain and frequently show lateral self-association in order to form complex connective tissues. Several collagens also play a role in cell adhesion, important for maintaining normal tissue architecture and function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL8A1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene COL8A1
Gene Accession No. P27658, Q00780
Gene Alias C3orf7; cell proliferation-inducing protein 41; Col8a1; Col8a-1; Collagen alpha-1(VIII) chain; collagen type VIII alpha 1; collagen VIII, alpha-1 polypeptide; collagen, type VIII, alpha 1; endothelial collagen; procollagen type VIII alpha 1; procollagen, type VIII, alpha 1; smag-64; smooth muscle cell-expressed and macrophage conditioned medium-induced protein smag-64; Vastatin
Gene Symbols COL8A1
Host Species Rabbit
Immunogen Recombinant Human Collagen alpha-1(VIII) chain protein (572-744AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12837, 1295
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.