Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COL8A2 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA562046
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562046 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562046 Supplier Thermo Scientific Supplier No. PA562046

Rabbit Polyclonal Antibody

Immunogen sequence: GYAPVKYIQP MQKGPVGPPF REGKGQYLEM PLPLLPMDLK G Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

This gene encodes the alpha 2 chain of type VIII collagen. The protein is a major component of the basement membrane of the corneal endothelium and forms homo- or heterotrimers with alpha 1 (VIII) type collagens. Defects in this gene are associated with Fuchs endothelial corneal dystrophy and posterior polymorphous corneal dystrophy type 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL8A2
Applications Immunohistochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COL8A2
Gene Accession No. P25067
Gene Alias AI429819; COL8A2; Collagen alpha-2(VIII) chain; collagen type VIII alpha 2; collagen VIII, alpha-2 polypeptide; collagen, type VIII, alpha 2; endothelial collagen; FECD; FECD1; PPCD; PPCD2; procollagen type VIII alpha 2; procollagen, type VIII, alpha 2
Gene Symbols COL8A2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COL8A2
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1296
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.