Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COL9A1 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299959PCP
Description
COL9A1 Polyclonal antibody specifically detects COL9A1 in Human samples. It is validated for ELISASpecifications
COL9A1 | |
Polyclonal | |
ELISA | |
alpha-1 polypeptide, collagen, type IX, alpha 1 | |
Produced in rabbits immunized with purified, recombinant Human COL9A1 (Accession#: P20849-3; Met1-Pro328) | |
0.1 mL | |
Cell Biology, Cytoskeleton Markers, Signal Transduction | |
1297 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction