Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COL9A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103380
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103380 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103380 Supplier Invitrogen™ Supplier No. PA5103380
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Collagen IX alpha 1.

COL9A1 is one of the three alpha chains of type IX collagen, which is a minor (5-20%) collagen component of hyaline cartilage. Type IX collagen is usually found in tissues containing type II collagen, a fibrillar collagen. Studies in knockout mice have shown that synthesis of the alpha 1 chain is essential for assembly of type IX collagen molecules, a heterotrimeric molecule, and that lack of type IX collagen is associated with early onset osteoarthritis. Mutations in the COL9A1 gene are associated with osteoarthritis in humans, with multiple epiphyseal dysplasia, 6, a form of chondrodysplasia, and with Stickler syndrome, a disease characterized by ophthalmic, orofacial, articular, and auditory defects.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL9A1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene COL9A1
Gene Accession No. P20849, Q05722
Gene Alias alpha 1 type IX collagen; alpha-1(IX) collagen chain; cartilage-specific short collagen; COL9A1; Col9a-1; Collagen alpha-1(IX) chain; collagen IX, alpha-1 polypeptide; collagen type IX alpha 1; collagen, type IX, alpha 1; DJ149L1.1.2; EDM6; MED; procollagen, type IX, alpha 1; STL4
Gene Symbols COL9A1
Host Species Rabbit
Immunogen A synthesized peptide derived from human COL9A1(Accession P20849), corresponding to amino acid residues G424-T474.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12839, 1297
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.