Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COL9A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563286
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563286 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563286 Supplier Invitrogen™ Supplier No. PA563286
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PGKPGRPGTI QGLEGSADFL CPTNCPPGMK GPPGLQGV Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

This gene encodes one of the three alpha chains of type IX collagen, the major collagen component of hyaline cartilage. Type IX collagen, a heterotrimeric molecule, is usually found in tissues containing type II collagen, a fibrillar collagen. This chain is unusual in that, unlike the other two type IX alpha chains, it contains a covalently attached glycosaminoglycan side chain. Mutations in this gene are associated with multiple epiphyseal dysplasia. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL9A2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COL9A2
Gene Accession No. Q14055
Gene Alias AI427499; alpha 1 type IX collagen; alpha 2 type IX collagen; COL9A2; Col9a-2; Collagen alpha-2(IX) chain; collagen IX, alpha-2 polypeptide; collagen type IX alpha 2; collagen, type IX, alpha 2; DJ39G22.4; EDM2; MED; procollagen, type IX, alpha 2; STL5
Gene Symbols COL9A2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COL9A2. Recombinant protein control fragment (Product #RP-100440).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1298
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.