Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COL9A3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | COL9A3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
COL9A3 Polyclonal specifically detects COL9A3 in Human samples. It is validated for Western Blot.Specifications
| COL9A3 | |
| Polyclonal | |
| Rabbit | |
| Q14050 | |
| 1299 | |
| Synthetic peptides corresponding to COL9A3 (collagen, type IX, alpha 3) The peptide sequence was selected from the C terminal of COL9A3. Peptide sequence GSKGEKGERGRAGELGEAGPSGEPGVPGDAGMPGERGEAGHRGSAGALGP. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| collagen type IX proteoglycan, collagen, type IX, alpha 3, DJ885L7.4.1, EDM3collagen alpha-3(IX) chain, FLJ90759, IDDcollagen IX, alpha-3 polypeptide, MED | |
| COL9A3 | |
| IgG | |
| 63 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title