Learn More
Invitrogen™ COL9A3 Polyclonal Antibody

Description
The antibody detects endogenous levels of total COL9A3 protein.
Specifications
Specifications
| Antigen | COL9A3 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | COL9A3 |
| Gene Accession No. | Q14050 |
| Gene Alias | alpha 3 type IX collagen; AV006866; cb367; CO9A3; COL9A3; Collagen alpha-3(IX) chain; collagen IX, alpha-3 polypeptide; collagen type IX alpha 3; collagen type IX alpha 3 chain; collagen type IX proteoglycan; collagen, type IX, alpha 3; DJ885L7.4.1; EDM3; IDD; MED; procollagen, type IX, alpha 3; sb:cb367; si:ch73-162j3.1; wu:fa04f01 |
| Gene Symbols | COL9A3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.