Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ COL9A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566159
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566159 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566159 Supplier Invitrogen™ Supplier No. PA566159
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PGLPGAIGAQG TPGICDTSAC QGAVLGGVGE KSGSRS Highest antigen sequence identity to the following orthologs - mouse 92%, rat 92%.

This gene encodes one of the three alpha chains of type IX collagen, the major collagen component of hyaline cartilage. Type IX collagen, a heterotrimeric molecule, is usually found in tissues containing type II collagen, a fibrillar collagen. Mutations in this gene are associated with multiple epiphyseal dysplasia type 3. [provided by RefSeq, Jan 2010].
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL9A3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COL9A3
Gene Accession No. Q14050
Gene Alias alpha 3 type IX collagen; AV006866; cb367; CO9A3; COL9A3; Collagen alpha-3(IX) chain; collagen IX, alpha-3 polypeptide; collagen type IX alpha 3; collagen type IX alpha 3 chain; collagen type IX proteoglycan; collagen, type IX, alpha 3; DJ885L7.4.1; EDM3; IDD; MED; procollagen, type IX, alpha 3; sb:cb367; si:ch73-162j3.1; wu:fa04f01
Gene Symbols COL9A3
Host Species Rabbit
Immunogen Recombinant Human COL9A3. Recombinant protein control fragment (Product #RP-106844).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1299
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.