Learn More
Thermo Scientific Chemicals Colchicine, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
- Colchicine (7alphah-colchicine) is an alkaloid found in Colchicum autumnale that has been shown to have anti-gout and anti-inflammatory properties
- Colchicine binds tubulin and prevents the migration of immune cells, reducing inflammation
Applications
- Colchicine is an anti-mitotic; may be used in the study of cell proliferation and migration
- Intended for in vivo and in vitro research applications
- For research use only, not intended to treat disease
Specifications
Specifications
| Melting Point | 150.0°C to 160.0°C |
| Color | Yellow |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% min. on anhydrous and solvent-free substance (HPLC) |
| Packaging | Glass bottle |
| Quantity | 5 g |
| Beilstein | 14,IV,946 |
| Merck Index | 15,2455 |
| Specific Rotation Condition | − 250.00 (20.00°C c=1,alcohol) |
| Specific Rotation | − 250.00 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement: Fatal if swallowed. May cause genetic defects. |
| Precautionary Statement | GHS P Statement: IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 200-598-5 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.