Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Colchicine
GREENER_CHOICESDP

Catalog No. 13641G Shop All Novus Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-641-G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 13-641-G Supplier R&D Systems™ Supplier No. 1364/1G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Inhibitor of tubulin

Plant-derived alkaloid that binds to tubulin and depolymerizes microtubules. Identified as a candidate for repurposing for COVID-19.

Chemical Identifiers

CAS 64-86-8
Molecular Formula C22H25NO6

Specifications

Quantity 1 g
Product Line Tocris
Formula Weight 399.44
Purity >98%
Molecular Weight (g/mol) 399.44
Chemical Name or Material Colchicine
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.