Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Cold Dissociation Enzyme Mix - Kidney

Catalog No. 09924464 Shop All Cytiva Products
Change view
Click to view available options
Cell Line:
Enzyme, Kidney
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cell Line
09-924-464 Enzyme, Kidney
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-924-464 Supplier Cytiva Supplier No. 29733434
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A standalone kit, intended for the dissociation of kidney tissue in single-cell sequencing.

  • Works effectively at 4°C
  • Minimized cell death and stress (defined as: apoptosis pathway markers, macrophages, etc.) and improved transcriptional profile of the single cells for more accurate results in single-cell sequencing and analysis
  • Ready-made solution specifically for kidney tissue
  • Optimized for the dissociation of the different segments that are operative in kidney tissue
  • Optimized for use with the VIA Extractor™ tissue disaggregator
  • One of the few systems that can sustain a cold temperature in a controlled manner. Effective dissociation at cooler temperatures increases viability and minimizes cell aggregation
TRUSTED_SUSTAINABILITY

Specifications

Cell Line Enzyme, Kidney
Cell Type Enzyme, Kidney
Culture Type Enzyme, Kidney
Form Liquid
Product Type Cell Culture Dissociation Reagent
Reagent Type Enzyme
Sterility Non-sterile
With Additives None
Without Additives None
Formulation Liquid Buffer and Enzyme Solution

For research use only (RUO). Not for diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.