Learn More
CPC™ HFC35 Polysulfone Hose Barb Coupling Inserts
Catalog No. p-4902201 Shop All Colder Products Company Products
Click to view available options
Product Type:
Hose Barb Non-valved In-line Coupling Insert
Hose Barb Valved In-line Coupling Insert
Hose Barb:
1/2 in.
3/8 in.
Free floating
  • Polysulfone couplings
  • Withstands demanding applications
  • UV resitant, able to withstand continued exposure to harmful UV rays
  • Ergonomic design
  • Operate in harsh environments in temperatures from -40°F to 280°F (-40°C to 138°C)
  • Pressure: Vacuum to 125psi (8.6 bar)
  • Large, shrouded thumb latch pad
  • Leads to high flow and exceptionally low spillage
Specifications
Material Polysulfone
Hose Barb 3/8 in.
Product Type Hose Barb Valved In-line Coupling Insert
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.