Description
- 36 tissue block capacity
- Designed to take up minimal bench-top space
- Removes the need for chilling sprays or ice blocks
- Remains cold for up to 4 hours
- No electrical components or compressors required
Specifications
Specifications
Product Type | Coldplate |
For Use With (Equipment) | Chilling Sprays, Ice Blocks |