Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COLEC10 Antibody (4F9D7), Novus Biologicals™
SDP

Catalog No. p-7200013 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP262807 100 μg
NB019781 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP262807 Supplier Novus Biologicals Supplier No. NBP262807
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

COLEC10 Monoclonal specifically detects COLEC10 in Human samples. It is validated for Western Blot, Immunoassay.

Specifications

Antigen COLEC10
Applications In Situ Hybridization (ISH), Western Blot
Classification Monoclonal
Clone 4F9D7
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunoassay
Formulation PBS with 0.1% BSA with 0.02% Sodium Azide
Gene Alias CL-34, CL-L1, CLL1collectin 34, collectin liver 1, Collectin liver protein 1, collectin sub-family member 10 (C-type lectin), collectin-10, collectin-34, MGC118794, MGC118795
Gene Symbols COLEC10
Host Species Mouse
Immunogen Collectin-10
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 10584
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.